2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

059875-1G 059875-5G
101919-380EA 58.46 USD
101919-380 102833-288
2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine ≥95%
2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
MDL Number: MFCD08059281
CAS Number: 63200-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR