4-Chloro-5H-pyrrolo[3,2-d]pyrimidine-7-carbaldehyde ≥96%

Supplier: Matrix Scientific

059873-100MG 059873-250MG
101919-370EA 218.46 USD
101919-370 101919-372
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine-7-carbaldehyde ≥96%
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine-7-carbaldehyde

Hazard: Irritant

Formula: C₇H₄ClN₃O
MDL Number: MFCD19105252
CAS Number: 1311275-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR