Ethyl-4-chloro-5H-pyrrolo[3,2-d]pyrimidine-7-carboxylate ≥96%

Supplier: Matrix Scientific

059870-250MG 059870-1G
101919-358EA 678.46 USD
101919-358 101919-360
Ethyl-4-chloro-5H-pyrrolo[3,2-d]pyrimidine-7-carboxylate ≥96%
Ethyl-4-chloro-5H-pyrrolo[3,2-d]pyrimidine-7-carboxylate

Hazard: Irritant

Formula: C₉H₈ClN₃O₂
MDL Number: MFCD10566079
CAS Number: 853058-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR