Ethyl-4-chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate

059869-250MG 059869-1G
101919-354EA 235.38 USD
101919-354 101919-356
Ethyl-4-chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate ≥95%
Ethyl-4-chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₉H₈ClN₃O₂
MW: 225.6300 g/mol
MDL Number: MFCD09839778
CAS Number: 187725-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR