6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

059868-250MG 059868-1G
101919-350EA 184.62 USD
101919-350 101919-352
6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine ≥95%
6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₆H₃BrClN₃
MDL Number: MFCD08276174
CAS Number: 784150-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR