4,6-Dichloro-5-fluoro-2-methylpyrimidine ≥96%

Supplier: Matrix Scientific

059863-250MG 059863-1G
101919-334EA 353.85 USD
101919-334 101919-336
4,6-Dichloro-5-fluoro-2-methylpyrimidine ≥96%
4,6-Dichloro-5-fluoro-2-methylpyrimidine

Hazard: Irritant

Formula: C₅H₃Cl₂FN₂
MW: 181.0000 g/mol
MDL Number: MFCD11520450
CAS Number: 105806-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR