4,6-Dichloro-5-fluoropyrimidine ≥96%

Supplier: Matrix Scientific

059861-1G 059861-5G
101919-328EA 53.85 USD
101919-328 102833-286
4,6-Dichloro-5-fluoropyrimidine ≥96%
4,6-Dichloro-5-fluoropyrimidine

Hazard: Corrosive/Lachrymator

Formula: C₄HCl₂FN₂
MW: 166.97 g/mol
Boiling Pt: 50-60°C/14mmHg
Flash Pt: 91°C
MDL Number: MFCD08056331
CAS Number: 213265-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR