6-Chloro-2-cyclopropylpyrimidin-4-amine ≥96%

Supplier: Matrix Scientific

059859-250MG 059859-1G
101919-318EA 287.69 USD
101919-318 101919-320
6-Chloro-2-cyclopropylpyrimidin-4-amine ≥96%
6-Chloro-2-cyclopropylpyrimidin-4-amine

Hazard: Irritant

Formula: C₇H₈ClN₃
MDL Number: MFCD19105247
CAS Number: 1311275-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR