N-Allyl-4,6-dichloropyrimidin-5-amine ≥96%
Supplier: Matrix Scientific
059856-250MG
059856-1G
101919-308EA
218.46
USD
101919-308
101919-310
N-Allyl-4,6-dichloropyrimidin-5-amine ≥96%
N-Allyl-4,6-dichloropyrimidin-5-amine
Hazard: Irritant
Formula:
C₇H₇Cl₂N₃ |
MDL Number:
MFCD19105245 CAS Number: 889669-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...