4,6-Dichloro-2-cyclopropylpyrimidine ≥98%

Supplier: Matrix Scientific

059853-1G 059853-5G
101919-302EA 129.23 USD
101919-302 102551-554
4,6-Dichloro-2-cyclopropylpyrimidine ≥98%
4,6-Dichloro-2-cyclopropylpyrimidine

Hazard: Irritant

Formula: C₇H₆Cl₂N₂
Melting Pt: 105…107 °C
MDL Number: MFCD11046809
CAS Number: 7043-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR