4,6-Dichloro-5-fluoropyrimidin-2-amine ≥96%

Supplier: Matrix Scientific

059851-250MG 059851-1G
101919-292EA 313.85 USD
101919-292 101919-294
4,6-Dichloro-5-fluoropyrimidin-2-amine ≥96%
4,6-Dichloro-5-fluoropyrimidin-2-amine

Hazard: Irritant

Formula: C₄H₂Cl₂FN₃
MDL Number: MFCD11870221
CAS Number: 15598-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR