4-Chloro-5-tosyl-5H-pyrrolo[3,2-d]pyrimidine ≥98%

Supplier: Matrix Scientific

059847-250MG 059847-1G
101919-276EA 327.69 USD
101919-276 101919-278
4-Chloro-5-tosyl-5H-pyrrolo[3,2-d]pyrimidine ≥98%
4-Chloro-5-tosyl-5H-pyrrolo[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₁₃H₁₀ClN₃O₂S
MDL Number: MFCD19105242
CAS Number: 1311275-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR