6-Amino-2-cyclopropylpyrimidin-4-ol ≥96%

Supplier: Matrix Scientific

059845-250MG 059845-1G
101919-268EA 172.31 USD
101919-268 101919-270
6-Amino-2-cyclopropylpyrimidin-4-ol ≥96%
6-Amino-2-cyclopropylpyrimidin-4-ol

Hazard: Irritant

Formula: C₇H₉N₃O
MDL Number: MFCD16988316
CAS Number: 954388-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR