2-Cyclopropylpyrimidine-4,6-diol ≥98%
Supplier: Matrix Scientific
059844-250MG
059844-1G
101919-264EA
515.38
USD
101919-264
101919-266
2-Cyclopropylpyrimidine-4,6-diol ≥98%
2-Cyclopropylpyrimidine-4,6-diol
Hazard: Irritant
Formula:
C₇H₈N₂O₂ |
MDL Number:
MFCD19105241 CAS Number: 7024-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...