5-Allyl-2-cyclopropylpyrimidine-4,6-diol ≥98%

Supplier: Matrix Scientific

059843-250MG 059843-1G
101919-260EA 250.77 USD
101919-260 101919-262
5-Allyl-2-cyclopropylpyrimidine-4,6-diol ≥98%
5-Allyl-2-cyclopropylpyrimidine-4,6-diol

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MDL Number: MFCD19105240
CAS Number: 1311275-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR