2-Amino-5-fluoropyrimidine-4,6-diol ≥95%

Supplier: Matrix Scientific

059840-250MG 059840-1G
101919-250EA 227.54 USD
101919-250 101919-252
2-Amino-5-fluoropyrimidine-4,6-diol ≥95%
2-Amino-5-fluoropyrimidine-4,6-diol

Hazard: Irritant

Formula: C₄H₄FN₃O₂
MDL Number: MFCD11870220
CAS Number: 669-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR