5-Fluoropyrimidine-4,6-diol ≥96%

Supplier: Matrix Scientific

059839-250MG 059839-1G
101919-246EA 607.69 USD
101919-246 101919-248
5-Fluoropyrimidine-4,6-diol ≥96%
5-Fluoropyrimidine-4,6-diol

Hazard: Irritant

Formula: C₄H₃FN₂O₂
MDL Number: MFCD09033260
CAS Number: 106615-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR