Ethyl-3,5-dichloro-4-cyanopyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

059809-500MG 059809-1G
101919-140EA 1192.31 USD
101919-140 101919-142
Ethyl-3,5-dichloro-4-cyanopyridine-2-carboxylate ≥95%
Ethyl-3,5-dichloro-4-cyanopyridine-2-carboxylate

Hazard: Irritant

Formula: C₉H₆Cl₂N₂O₂
MW: 245.07 g/mol
MDL Number: MFCD18904316
CAS Number: 1296172-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR