5-(Diethoxymethyl)-2-fluorobenzonitrile ≥95%

Supplier: Matrix Scientific

059807-500MG 059807-1G 059807-5G
101919-130EA 298.46 USD
101919-130 101919-132 101919-134
5-(Diethoxymethyl)-2-fluorobenzonitrile ≥95%
5-(Diethoxymethyl)-2-fluorobenzonitrile

Hazard: Irritant

Formula: C₁₂H₁₄FNO₂
MDL Number: MFCD18904305
CAS Number: 1176161-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR