Benzyl 4-[(3-{[(tert-butoxy)carbonyl]amino}propyl)amino]piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

059801-500MG 059801-1G 059801-5G
101919-104EA 298.46 USD
101919-104 101919-106 101919-108
Benzyl 4-[(3-{[(tert-butoxy)carbonyl]amino}propyl)amino]piperidine-1-carboxylate ≥95%
Benzyl 4-[(3-{[(tert-butoxy)carbonyl]amino}propyl)amino]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₂₁H₃₃N₃O₄
MDL Number: MFCD18904302
CAS Number: 936129-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR