Benzyl-4-[(2-{[(tert-butoxy)carbonyl]amino}ethyl)amino]piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

059800-500MG 059800-1G 059800-5G
101919-098EA 298.46 USD
101919-098 101919-100 101919-102
Benzyl-4-[(2-{[(tert-butoxy)carbonyl]amino}ethyl)amino]piperidine-1-carboxylate ≥95%
Benzyl-4-[(2-{[(tert-butoxy)carbonyl]amino}ethyl)amino]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₂₀H₃₁N₃O₄
MW: 377.49 g/mol
MDL Number: MFCD18904301
CAS Number: 864293-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR