1-[(2-Aminoethyl)sulfanyl]-4-bromobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(4-Bromophenyl)sulphanyl]ethan-1-amine

059795-500MG 059795-1G
101919-078EA 298.46 USD
101919-078 101919-080
1-[(2-Aminoethyl)sulfanyl]-4-bromobenzene ≥95%
1-[(2-Aminoethyl)sulfanyl]-4-bromobenzene

Hazard: Irritant

Formula: C₈H₁₀BrNS
MDL Number: MFCD05262934
CAS Number: 69326-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR