2,6-Difluoronicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Difluoropyridine-3-carbaldehyde, 2,6-Difluoropyridine-3-carboxaldehyde

059785-1G 059785-5G
101919-072EA 26.15 USD
101919-072 101919-074
2,6-Difluoronicotinaldehyde ≥95%
2,6-Difluoronicotinaldehyde

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₆H₃F₂NO
MW: 143.093 g/mol
MDL Number: MFCD08277633
CAS Number: 155601-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR