1-(Dimethylamino)-4,4-dimethylpent-1-en-3-one ≥95%

Supplier: Matrix Scientific

059639-500MG 059639-1G
101918-718EA 600 USD
101918-718 101918-720
1-(Dimethylamino)-4,4-dimethylpent-1-en-3-one ≥95%
1-(Dimethylamino)-4,4-dimethylpent-1-en-3-one

Hazard: Irritant

Formula: C₉H₁₇NO
MDL Number: MFCD11847681
CAS Number: 6135-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR