5-Chlorosulfonyl-2-methoxybenzoic acid ≥97%

Supplier: Matrix Scientific

059604-1G 059604-5G
101918-622EA 67.69 USD
101918-622 101918-624
5-Chlorosulfonyl-2-methoxybenzoic acid ≥97%
5-Chlorosulfonyl-2-methoxybenzoic acid

Hazard: Irritant

Formula: C₈H₇ClO₅S
MDL Number: MFCD00066912
CAS Number: 51904-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR