Methyl-1-(2-cyanophenyl)-5-methyl-1H-pyrazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

059587-500MG 059587-1G 059587-5G
101918-562EA 600 USD
101918-562 101918-564 101918-566
Methyl-1-(2-cyanophenyl)-5-methyl-1H-pyrazole-3-carboxylate ≥95%
Methyl-1-(2-cyanophenyl)-5-methyl-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₁N₃O₂
MDL Number: MFCD18651786
CAS Number: 1272756-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR