Ethyl (2Z)-2-cyano-3-hydroxy-3-phenylprop-2-enoate ≥95%

Supplier: Matrix Scientific

059573-500MG 059573-1G 059573-5G
101918-496EA 193.85 USD
101918-496 101918-498 101918-500
Ethyl (2Z)-2-cyano-3-hydroxy-3-phenylprop-2-enoate ≥95%
Ethyl (2Z)-2-cyano-3-hydroxy-3-phenylprop-2-enoate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD00995909
CAS Number: 59551-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR