Ethyl (2Z)-2-cyano-3-hydroxypent-2-enoate ≥95%

Supplier: Matrix Scientific

059572-500MG 059572-1G
101918-492EA 193.85 USD
101918-492 101918-494
Ethyl (2Z)-2-cyano-3-hydroxypent-2-enoate ≥95%
Ethyl (2Z)-2-cyano-3-hydroxypent-2-enoate

Hazard: Irritant

Formula: C₈H₁₁NO₃
MDL Number: MFCD18651766
CAS Number: 1273562-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR