Ethyl-1-[(2-aminophenyl)carbonyl]piperidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

059562-500MG 059562-1G
101918-442EA 193.85 USD
101918-442 101918-444
Ethyl-1-[(2-aminophenyl)carbonyl]piperidine-3-carboxylate ≥95%
Ethyl-1-[(2-aminophenyl)carbonyl]piperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₃
MDL Number: MFCD12560018
CAS Number: 1039964-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR