2-(2-Chloroethyl)-2H,3H,4H-pyrido-[3,2-b][1,4]oxazin-3-one ≥95%

Supplier: Matrix Scientific

059553-500MG 059553-1G
101918-408EA 600 USD
101918-408 101918-410
2-(2-Chloroethyl)-2H,3H,4H-pyrido-[3,2-b][1,4]oxazin-3-one ≥95%
2-(2-Chloroethyl)-2H,3H,4H-pyrido-[3,2-b][1,4]oxazin-3-one

Hazard: Irritant

Formula: C₉H₉ClN₂O₂
MDL Number: MFCD04125675
CAS Number: 866040-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR