2-(3-Bromopropyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one ≥95%
Supplier: Matrix Scientific
059551-500MG
059551-1G
101918-402EA
600
USD
101918-402
101918-404
2-(3-Bromopropyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one ≥95%
2-(3-Bromopropyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one
Hazard: Irritant
Formula:
C₁₀H₁₁BrN₂O₂ |
MDL Number:
MFCD18651758 CAS Number: 1272756-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...