2-(2-Bromoethyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one ≥95%

Supplier: Matrix Scientific

059548-500MG 059548-1G
101918-392EA 600 USD
101918-392 101918-394
2-(2-Bromoethyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one ≥95%
2-(2-Bromoethyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one

Hazard: Irritant

Formula: C₉H₉BrN₂O₂
MDL Number: MFCD18651760
CAS Number: 1272756-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR