2-(4-Bromobutyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one ≥95%
Supplier: Matrix Scientific
059547-500MG
059547-1G
101918-388EA
600
USD
101918-388
101918-390
2-(4-Bromobutyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one ≥95%
2-(4-Bromobutyl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one
Hazard: Irritant
Formula:
C₁₁H₁₃BrN₂O₂ |
MDL Number:
MFCD18651759 CAS Number: 1272756-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...