Ethyl-trans-3-(3-pyridyl)acrylate ≥95%

Supplier: Matrix Scientific

054890-500MG 054890-1G 054890-5G
101908-938EA 76.92 USD
101908-938 101908-940 101908-942
Ethyl-trans-3-(3-pyridyl)acrylate ≥95%
Ethyl-trans-3-(3-pyridyl)acrylate

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.20 g/mol
MDL Number: MFCD02177443
CAS Number: 59607-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR