Ethyl (E)-3-(2-pyridinyl)-2-propenoate ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-Ethyl 3-(pyridin-2-yl)acrylate

054889-500MG 054889-1G 054889-5G
101908-932EA 181.54 USD
101908-932 101908-934 101908-936
Ethyl (E)-3-(2-pyridinyl)-2-propenoate ≥95%
Ethyl (E)-3-(2-pyridinyl)-2-propenoate

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
MDL Number: MFCD16140351
CAS Number: 70526-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR