Ethyl-2-((4-methoxybenzyl)amino)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-((4-methoxybenzyl)amino)acetate

054887-500MG 054887-1G 054887-5G
101908-922EA 146.15 USD
101908-922 101908-924 101908-926
Ethyl-2-((4-methoxybenzyl)amino)acetate ≥95%
Ethyl-2-((4-methoxybenzyl)amino)acetate

Hazard: Irritant

Formula: C₁₂H₁₇NO₃
MW: 223.2700 g/mol
MDL Number: MFCD11151999
CAS Number: 60857-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR