[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (1R,5S,6R)-3-Aza-3-benzyl-6-(hydroxymethyl)bicyclo[3.1.0]hexane, rel-((1R,5S,6r)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl)methanol

054862-500MG 054862-1G
101908-828EA 333.85 USD
101908-828 101908-830
[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol ≥95%
[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol

Hazard: Irritant

Formula: C₁₃H₁₇NO
MW: 203.29 g/mol
MDL Number: MFCD16140332
CAS Number: 134575-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR