1H-1,2,3-Triazole-1-acetic acid hydrochloride ≥95%

Supplier: Matrix Scientific

054853-500MG 054853-1G
101908-792EA 556.92 USD
101908-792 101908-794
1H-1,2,3-Triazole-1-acetic acid hydrochloride ≥95%
1H-1,2,3-Triazole-1-acetic acid hydrochloride

Hazard: Irritant

Formula: C₄H₆ClN₃O₂
MW: 163.56 g/mol
MDL Number: MFCD09029581
CAS Number: 4314-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR