Thiazol-5-ylmethanamine ≥95%

Supplier: Matrix Scientific

054852-500MG 054852-1G
101908-788EA 556.92 USD
101908-788 101908-790
Thiazol-5-ylmethanamine ≥95%
Thiazol-5-ylmethanamine

Hazard: Irritant

Formula: C₄H₆N₂S
CAS Number: 161805-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR