tert-Butyl-3-(hydrazinocarbonyl)tetrahydro-1(2H)-pyridine carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(hydrazinecarbonyl)piperidine-1-carboxylate

054844-1G 054844-5G
101908-760EA 121.54 USD
101908-760 101908-762
tert-Butyl-3-(hydrazinocarbonyl)tetrahydro-1(2H)-pyridine carboxylate ≥97%
tert-Butyl-3-(hydrazinocarbonyl)tetrahydro-1(2H)-pyridine carboxylate

Hazard: Irritant

Formula: C₁₁H₂₁N₃O₃
MW: 243.31 g/mol
MDL Number: MFCD06795929
CAS Number: 625470-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR