4-Spiro[1-thiophthalane]piperidine hydrochloride ≥95%

Supplier: Matrix Scientific

054840-500MG 054840-1G
101908-744EA 744.92 USD
101908-744 101908-746
4-Spiro[1-thiophthalane]piperidine hydrochloride ≥95%
4-Spiro[1-thiophthalane]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClNS
MW: 241.79 g/mol
MDL Number: MFCD02179150
CAS Number: 191673-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR