4-Spiro[3-phthalide]piperidine hydrochloride ≥95%

Supplier: Matrix Scientific

054839-500MG 054839-1G
101908-740EA 1109.23 USD
101908-740 101908-742
4-Spiro[3-phthalide]piperidine hydrochloride ≥95%
4-Spiro[3-phthalide]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₂
MW: 239.7 g/mol
MDL Number: MFCD02179149
CAS Number: 172733-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR