4-Spiroindanepiperidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydrospiro[indene-1,4'-piperidine] hydrochloride

054836-500MG 054836-1G
101908-728EA 460 USD
101908-728 101908-730
4-Spiroindanepiperidine hydrochloride ≥95%
4-Spiroindanepiperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClN
MW: 223.75 g/mol
MDL Number: MFCD08461061
CAS Number: 96651-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR