2-(Pyridin-4-ylmethoxy)acetic acid hydrochloride hydrate ≥95%

Supplier: Matrix Scientific

054832-500MG 054832-1G
101908-712EA 921.54 USD
101908-712 101908-714
2-(Pyridin-4-ylmethoxy)acetic acid hydrochloride hydrate ≥95%
2-(Pyridin-4-ylmethoxy)acetic acid hydrochloride hydrate

Hazard: Irritant

Formula: C₈H₁₂ClNO₄
MDL Number: MFCD16090036
CAS Number: 1452518-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR