1-(4-Piperidyl)-1, 2, 3, 4-tetrahydro-2-quinolinone hydrochloride ≥95%

Supplier: Matrix Scientific

054830-500MG 054830-1G
101908-704EA 723.08 USD
101908-704 101908-706
1-(4-Piperidyl)-1, 2, 3, 4-tetrahydro-2-quinolinone hydrochloride ≥95%
1-(4-Piperidyl)-1, 2, 3, 4-tetrahydro-2-quinolinone hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O
MDL Number: MFCD16090018
CAS Number: 141134-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR