1-(4-Piperidyl)-1, 2-dihydro-2-quinolinone ≥95%

Supplier: Matrix Scientific

054829-500MG 054829-1G
101908-700EA 692.31 USD
101908-700 101908-702
1-(4-Piperidyl)-1, 2-dihydro-2-quinolinone ≥95%
1-(4-Piperidyl)-1, 2-dihydro-2-quinolinone

Hazard: Irritant

Formula: C₁₄H₁₇ClN₂O
MDL Number: MFCD16090017
CAS Number: 173843-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR