1-(Piperidin-4-yl)-1H-benzo[d][1,3]oxazin-2(4H)-one hydrochloride ≥95%

Supplier: Matrix Scientific

054827-500MG 054827-1G
101908-692EA 1500 USD
101908-692 101908-694
1-(Piperidin-4-yl)-1H-benzo[d][1,3]oxazin-2(4H)-one hydrochloride ≥95%
1-(Piperidin-4-yl)-1H-benzo[d][1,3]oxazin-2(4H)-one hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O₂
MDL Number: MFCD09999158
CAS Number: 162045-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR