4-Oxo-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

054824-500MG 054824-1G
101908-680EA 750.77 USD
101908-680 101908-682
4-Oxo-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid ≥95%
4-Oxo-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀O₃
MDL Number: MFCD00177946
CAS Number: 3123-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR