4-N,N-Dimethylcarbamoyloxybenzoic acid ≥95%

Supplier: Matrix Scientific

054819-500MG 054819-1G
101908-660EA 375.38 USD
101908-660 101908-662
4-N,N-Dimethylcarbamoyloxybenzoic acid ≥95%
4-N,N-Dimethylcarbamoyloxybenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
MDL Number: MFCD16090021
CAS Number: 1204296-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR