2-[2-[(tert-Butoxycarbonyl)amino]thiazol-4-yl]acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: N-BOC-2-amino-4-thiazolacetic acid, 2-(2-((tert-Butoxycarbonyl)amino)thiazol-4-yl)acetic acid, [2-[[[(2-Methyl-2-propanyl)oxy]carbonyl]amino]-1,3-thiazol-4-yl]acetic acid

054818-500MG 054818-1G
101908-656EA 556.92 USD
101908-656 101908-658
2-[2-[(tert-Butoxycarbonyl)amino]thiazol-4-yl]acetic acid ≥95%
2-[2-[(tert-Butoxycarbonyl)amino]thiazol-4-yl]acetic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₄S
MW: 258.29 g/mol
MDL Number: MFCD02682394
CAS Number: 89336-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR